Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diaminoanthraquinone 90.0+%, TCI America™
SDP

Catalog No. D353925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D353925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D353925G Supplier TCI America Supplier No. D353925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 128-95-0
Melting Point 270°C
Color Black-Purple
Molecular Formula C14H10N2O2
Quantity 25 g
MDL Number MFCD00001224
Synonym 1,4-diaminoanthraquinone, disperse violet 1, krisolamine, 9,10-anthracenedione, 1,4-diamino, grasol violet r, seacyl violet r, acetate red violet r, 1,4-diamino anthraquinone, setacyl violet r, oil violet r
InChI Key FBMQNRKSAWNXBT-UHFFFAOYSA-N
SMILES NC1=CC=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O
IUPAC Name 1,4-diamino-9,10-dihydroanthracene-9,10-dione
Molecular Weight (g/mol) 238.25
PubChem CID 31420
Formula Weight 238.25
Percent Purity ≥90.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diaminoanthraquinone
Show More Show Less
EINECSNumber (4)-0710&(5)-2540
RTECSNumber CB6300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.