Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diaminoanthraquinone, tech. 90%, Thermo Scientific™

Catalog No. AAA1915614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1915614 25 g
AAA1915622 100 g
AAA1915636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1915614 Supplier Thermo Scientific Chemicals Supplier No. A1915614

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Use as the intermediate of synthetic dyes. Use anthraquinone reductive dyes and dispersal dyes and acid dyes and intermediates. Itself to disperse dyestuffs purple. 1,4-Diaminoanthraquinone oxidized with different metal atoms gives rise to conducting coordination polymers consisting of difunctional ligand molecules linked by metal atoms. 1, 4-diaminoanthraquinone has solar cell applications. A hybrid Si/poly (1, 4-diaminoanthraquinone) photoconductive diode for optical sensor applications. Acts as the sequential incorporation of the mediator in electrochemical sensing of glucose on a gold electrode.

Solubility
Soluble in benzene, pyridine, the difficulties, slightly soluble in hot aniline acetate, ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-95-0
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.25
MDL Number MFCD00001224
InChI Key FBMQNRKSAWNXBT-UHFFFAOYSA-N
Synonym 1,4-diaminoanthraquinone, disperse violet 1, krisolamine, 9,10-anthracenedione, 1,4-diamino, grasol violet r, seacyl violet r, acetate red violet r, 1,4-diamino anthraquinone, setacyl violet r, oil violet r
PubChem CID 31420
IUPAC Name 1,4-diaminoanthracene-9,10-dione
SMILES NC1=CC=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O

Specifications

Melting Point 260°C to 265°C
Odor Odorless
Quantity 25 g
Beilstein 2216556
Solubility Information Soluble in benzene,pyridine,the difficulties,slightly soluble in hot aniline acetate,ethanol.
Formula Weight 238.25
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,4-Diaminoanthraquinone
Hazard Category Danger
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-922-6
RTECSNumber CB6300000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.