Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Diazabicyclo[2.2.2]Octane Bis(Sulfur Dioxide) Adduct

Catalog No. 7429375G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-293-75G 5 g
74-293-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 74-293-75G Supplier Sigma Aldrich Supplier No. 7429375G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) (DABSO) is a charge transfer complex that can be used as a sulfur dioxide surrogate:In palladium-catalyzed aminosulfonylation process.In reaction with aryl bromides to synthesize sodium aryl sulfinates.It can also be used to activate DMSO and o-vinylanilines for the synthesis of N-aryl-1H-benzo[d]imidazol-1-amine and 4-aryl quinolines, respectively.

Specifications

CAS 119752-83-9
Molecular Formula C6H12N2O4S2
Linear Formula C6H12N2O4S2
MDL Number MFCD22377802
Quantity 5 g
Synonym DABCO.(SO2)2; DABSO
Molecular Weight (g/mol) 240.3 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.