Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-diazoniabicyclo[2.2.2]octane, 1,4-disulfino-, bis(inner salt) | 119752-83-9 | MFCD22377802 | 97.0% | 240.31 | C6H12N2O4S2 | 25 G
SDP

Catalog No. 5000302215
Encompass_Preferred

DABCO-Bis(sulfur dioxide) (DABSO) is a stable crystalline adduct of 1,4-diazabicyclo[2.2.2]octane and sulfur dioxide that provides a convenient, easy-to-handle solid source of SO2 for laboratory organic synthesis. It releases sulfur dioxide under reaction conditions and is commonly used in sulfonylation, sulfonamide, and sulfone formation.

  • Solid, easy-to-weigh source of sulfur dioxide.
  • Stable, safer alternative to handling gaseous SO2.
  • High purity (97.0%) suitable for synthetic applications.
  • White to off-white solid for simple storage and handling.
  • Applicable to sulfonylation, sulfonamide, and sulfone syntheses.
  • Molecular formula C6H12N2O4S2; molecular weight ~240.31 g/mol.

Catalog No. 50-003-02215 Supplier Medchemexpress LLC Supplier No. 25GHYW033049
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DABCO-Bis(sulfur dioxide) (DABSO) is a stable crystalline adduct of 1,4-diazabicyclo[2.2.2]octane and sulfur dioxide that provides a convenient, easy-to-handle solid source of SO2 for laboratory organic synthesis. It releases sulfur dioxide under reaction conditions and is commonly used in sulfonylation, sulfonamide, and sulfone formation.

  • Solid, easy-to-weigh source of sulfur dioxide.
  • Stable, safer alternative to handling gaseous SO2.
  • High purity (97.0%) suitable for synthetic applications.
  • White to off-white solid for simple storage and handling.
  • Applicable to sulfonylation, sulfonamide, and sulfone syntheses.
  • Molecular formula C6H12N2O4S2; molecular weight ~240.31 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.