Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromo-2,5-dimethylbenzene, 98+%, Thermo Scientific™

Catalog No. AC279011000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279011000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC279011000 Supplier Thermo Scientific Chemicals Supplier No. 279011000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-24-4
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.96
MDL Number MFCD00000091
InChI Key QENIALCDPFDFHX-UHFFFAOYSA-N
Synonym 2,5-dibromo-p-xylene, benzene, 1,4-dibromo-2,5-dimethyl, p-xylene, 2,5-dibromo, 2,5-dibromo-1,4-dimethylbenzene, 2,5-dimethyl-1,4-dibromobenzene, 2,5-dibromo-4-xylene, 2,4-dibromobenzene, p-xylene,5-dibromo, pubchem14829, acmc-1c2tg
PubChem CID 66175
IUPAC Name 1,4-dibromo-2,5-dimethylbenzene
SMILES CC1=CC(Br)=C(C)C=C1Br

Specifications

Melting Point 72.0°C to 75.0°C
Quantity 100 g
Refractive Index 1.5430 to 1.5450
Formula Weight 263.95
Chemical Name or Material 1,4-Dibromo-2,5-dimethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.