Learn More
Medchemexpress LLC 1,4-dibromo-butane-1,1,2,2,3,3,4,4-d8 | 68375-92-8 | MFCD00084134 | 99.9% | 223.96 | C4D8Br2 | 500 MG
SDP
Catalog No. 5000280761
Encompass_Preferred
1,4-Dibromo-butane-d8 is the deuterium-labeled analogue of 1,4-dibromobutane designed for use as an isotopic tracer and internal standard in analytical chemistry. It produces predictable mass shifts that facilitate accurate quantification and tracking in structural, metabolic, and pharmacokinetic studies.
  • Deuterium-labeled analog for isotopic tracing.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity (99.9%).
  • Molecular formula C4D8Br2; molecular weight 223.96.
  • Available in small pack sizes for analytical workflows.
  • Useful for quantitative and metabolic investigations.
Catalog No. 50-002-80761 Supplier Medchemexpress LLC Supplier No. HYW242724500MG
1,4-Dibromo-butane-d8 is the deuterium-labeled analogue of 1,4-dibromobutane designed for use as an isotopic tracer and internal standard in analytical chemistry. It produces predictable mass shifts that facilitate accurate quantification and tracking in structural, metabolic, and pharmacokinetic studies.
  • Deuterium-labeled analog for isotopic tracing.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity (99.9%).
  • Molecular formula C4D8Br2; molecular weight 223.96.
  • Available in small pack sizes for analytical workflows.
  • Useful for quantitative and metabolic investigations.