Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-dibromo-butane-1,1,2,2,3,3,4,4-d8 | 68375-92-8 | MFCD00084134 | 99.9% | 223.96 g/mol | C4D8Br2 | 1 G
SDP

Catalog No. 5000280490
Encompass_Preferred

1,4-Dibromo-butane-1,1,2,2,3,3,4,4-d8 is the fully deuterated isotopologue of 1,4-dibromobutane used as a stable-isotope reagent and internal standard in research chemistry. It is offered as a high-purity, documented reagent with defined storage and solubility properties for analytical and synthetic workflows.

  • Fully deuterated isotopologue for isotope-labeling and tracer studies.
  • High chemical purity (99.91%) suitable for analytical applications.
  • Molecular weight 223.96 g/mol and formula C4D8Br2.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Storage stability specified for pure form and in solvent to maintain integrity.
  • Supplied with COA and SDS for quality and safety documentation.

Catalog No. 50-002-80490 Supplier Medchemexpress LLC Supplier No. HYW2427241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-Dibromo-butane-1,1,2,2,3,3,4,4-d8 is the fully deuterated isotopologue of 1,4-dibromobutane used as a stable-isotope reagent and internal standard in research chemistry. It is offered as a high-purity, documented reagent with defined storage and solubility properties for analytical and synthetic workflows.

  • Fully deuterated isotopologue for isotope-labeling and tracer studies.
  • High chemical purity (99.91%) suitable for analytical applications.
  • Molecular weight 223.96 g/mol and formula C4D8Br2.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Storage stability specified for pure form and in solvent to maintain integrity.
  • Supplied with COA and SDS for quality and safety documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.