Learn More
Medchemexpress LLC 1,4-Dibromobenzene-d4 | 4165-56-4 | 99.9% | C6D4Br2 | 1 G
SDP
Catalog No. 5000434671
Encompass_Preferred
1,4-Dibromobenzene-d4 is a deuterium-labeled compound primarily used as a tracer and internal standard for quantitative analysis in various analytical methods such as NMR, GC-MS, and LC-MS. Deuteration of compounds is studied for its potential to affect pharmacokinetic and metabolic profiles of drugs. The unlabeled compound, 1,4-Dibromobenzene, serves as a fumigant and chemical intermediate in pharmaceuticals.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for use with NMR, GC-MS, or LC-MS
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34671 Supplier Medchemexpress LLC Supplier No. HY76025S1G
1,4-Dibromobenzene-d4 is a deuterium-labeled compound primarily used as a tracer and internal standard for quantitative analysis in various analytical methods such as NMR, GC-MS, and LC-MS. Deuteration of compounds is studied for its potential to affect pharmacokinetic and metabolic profiles of drugs. The unlabeled compound, 1,4-Dibromobenzene, serves as a fumigant and chemical intermediate in pharmaceuticals.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for use with NMR, GC-MS, or LC-MS
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs