Learn More
Medchemexpress LLC 1,4-Dibromobenzene-d4 | 4165-56-4 | 99.9% | C6D4Br2 | 5 G
SDP
Catalog No. 5000433863
Encompass_Preferred
1,4-Dibromobenzene-d4 is a deuterium labeled 1,4-Dibromobenzene, used as a fumigant and chemical intermediate for pharmaceuticals. This compound is for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-33863 Supplier Medchemexpress LLC Supplier No. HY76025S5G
1,4-Dibromobenzene-d4 is a deuterium labeled 1,4-Dibromobenzene, used as a fumigant and chemical intermediate for pharmaceuticals. This compound is for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs