Learn More
Cayman Chemical 1,4-Dicaffeoylquinic Acid, Purity: ≥98%, CAS: 1182-34-9, 10 mg
SDP
Catalog No. 502948456
Encompass_Preferred
A polyphenol with diverse biological activities; scavenges DPPH radicals; inhibits XO (IC50 = 7.36 µM); decreases protein levels of tyrosinase and MITF in B16/F1 murine melanocytes, inhibits melanogenesis in the same cells, and decreases tyrosinase activity in cell lysates at 25 µM; increases biliary flow in rats
Catalog No. 50-294-8456 Supplier Cayman Chemical Supplier No. 3384210 MG
A polyphenol with diverse biological activities; scavenges DPPH radicals; inhibits XO (IC50 = 7.36 µM); decreases protein levels of tyrosinase and MITF in B16/F1 murine melanocytes, inhibits melanogenesis in the same cells, and decreases tyrosinase activity in cell lysates at 25 µM; increases biliary flow in rats