Learn More
Medchemexpress LLC 1,4-Dicaffeoylquinic acid | 1182-34-9 | 516.45 | 5 MG
SDP
Catalog No. 5000389645
Encompass_Preferred
1,4-Dicaffeoylquinic acid (1,4-DCQA) is a phenylpropanoid compound isolated from Xanthii fructus. It acts as an inhibitor of xanthine oxidase with an IC50 of 7.36 μM and exhibits anti-inflammatory activity by inhibiting the production of TNF-α induced by LPS.
  • Isolated from Xanthii fructus.
  • Inhibitor of xanthine oxidase (IC50: 7.36 μM).
  • Possesses anti-inflammatory activity.
  • Inhibits the production of TNF-α induced by LPS.
  • No cytotoxicity observed in RAW 264.7 cells at concentrations up to 50 μM.
  • Purity: 99.25%.
Catalog No. 50-003-89645 Supplier Medchemexpress LLC Supplier No. HYN03585MG
1,4-Dicaffeoylquinic acid (1,4-DCQA) is a phenylpropanoid compound isolated from Xanthii fructus. It acts as an inhibitor of xanthine oxidase with an IC50 of 7.36 μM and exhibits anti-inflammatory activity by inhibiting the production of TNF-α induced by LPS.
  • Isolated from Xanthii fructus.
  • Inhibitor of xanthine oxidase (IC50: 7.36 μM).
  • Possesses anti-inflammatory activity.
  • Inhibits the production of TNF-α induced by LPS.
  • No cytotoxicity observed in RAW 264.7 cells at concentrations up to 50 μM.
  • Purity: 99.25%.