Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dichloro-2,5-dimethoxybenzene, TCI America™
SDP

Catalog No. D326625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D326625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D326625G Supplier TCI America Supplier No. D326625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2675-77-6
Molecular Formula C8H8Cl2O2
Molecular Weight (g/mol) 207.05
MDL Number MFCD00055445
InChI Key PFIADAMVCJPXSF-UHFFFAOYSA-N
Synonym chloroneb, demosan, tersan sp, demasan, tersan-sp, chloronebe, demosan 65w, terraneb, terraneb sp, soil fungicide 1823
PubChem CID 17581
ChEBI CHEBI:3628
IUPAC Name 1,4-dichloro-2,5-dimethoxybenzene
SMILES COC1=CC(Cl)=C(OC)C=C1Cl

Specifications

Melting Point 131°C
Color Yellow
Boiling Point 262°C
Quantity 25 g
UN Number 3077
Formula Weight 207.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dichloro-2,5-dimethoxybenzene
EINECSNumber (3)-0955
RTECSNumber CZ4750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.