Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dichloro-2-nitrobenzene, 98%

Catalog No. p-7081763
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1189636 500 g
AAA118960E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1189636 Supplier Thermo Scientific Chemicals Supplier No. A1189636
Only null left
Add to Cart

CAS: 89-61-2 | C6H3Cl2NO2 | 192.00 g/mol

1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-61-2
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007074
InChI Key RZKKOBGFCAHLCZ-UHFFFAOYSA-N
Synonym 2,5-dichloronitrobenzene, nitro-p-dichlorobenzene, benzene, 1,4-dichloro-2-nitro, 1-nitro-2,5-dichlorobenzene, 2,5-dichlornitrobenzen, 2,5-dichloro-1-nitrobenzene, benzene, 2,5-dichloronitro, 2,5-dichloronitrobenzol, unii-iy18g50ff4, ccris 3096
PubChem CID 6977
IUPAC Name 1,4-dichloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=CC=C1Cl

Specifications

Melting Point 53°C to 57°C
Density 1.6
Boiling Point 266°C to 269°C
Flash Point 125°C (257°F)
Quantity 500 g
UN Number UN3077
Beilstein 778109
Solubility Information Soluble in water,ethanol,ether,benzene,carbon disulfide. Slightly soluble in carbon tetrachloride.
Formula Weight 192
Percent Purity 98%
Chemical Name or Material 1,4-Dichloro-2-nitrobenzene
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H341-H302-H319
Suspected of causing genetic defects.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 201-923-3
RTECSNumber CZ5260000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.