Learn More
Medchemexpress LLC 1,4-dichlorobenzene | 106-46-7 | MFCD00000604 | 147.00 g·mol⁻¹ | C6H4Cl2 | 10g
SDP
Catalog No. 5000269970
Encompass_Preferred
1 4-Dichlorobenzene is used as an intermediate product in the manufacture of pigments pesticides and disinfectants 1 4-Dichlorobenzene is also employed as a moth control agent[1]
Catalog No. 50-002-69970 Supplier Medchemexpress LLC Supplier No. HYY049610G
1 4-Dichlorobenzene is used as an intermediate product in the manufacture of pigments pesticides and disinfectants 1 4-Dichlorobenzene is also employed as a moth control agent[1]