Learn More
Medchemexpress LLC 1,4-dichlorobenzene | 106-46-7 | MFCD00000604 | 147.00 g/mol | C6H4Cl2 | 1g
SDP
Catalog No. 5000268490
Encompass_Preferred
1 4-Dichlorobenzene is used as an intermediate product in the manufacture of pigments pesticides and disinfectants 1 4-Dichlorobenzene is also employed as a moth control agent[1]
Catalog No. 50-002-68490 Supplier Medchemexpress LLC Supplier No. HYY04961G
1 4-Dichlorobenzene is used as an intermediate product in the manufacture of pigments pesticides and disinfectants 1 4-Dichlorobenzene is also employed as a moth control agent[1]