Learn More
Medchemexpress LLC 1,4-dichlorobenzene | 106-46-7 | MFCD00000604 | 99.7% | 147.00 | C6H4Cl2 | 25g
SDP
Catalog No. 5000268172
Encompass_Preferred
1 4-Dichlorobenzene is used as an intermediate product in the manufacture of pigments pesticides and disinfectants 1 4-Dichlorobenzene is also employed as a moth control agent[1]
Catalog No. 50-002-68172 Supplier Medchemexpress LLC Supplier No. HYY049625G
1 4-Dichlorobenzene is used as an intermediate product in the manufacture of pigments pesticides and disinfectants 1 4-Dichlorobenzene is also employed as a moth control agent[1]