Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Dichlorobenzene

Catalog No. 501860387
Encompass
Change view
Click to view available options
Quantity:
100 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-186-0387 100 g
50-186-0388 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-186-0387 Supplier Sigma Aldrich Supplier No. D56829100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Dichlorobenzene
TRUSTED_SUSTAINABILITY

Specifications

CAS 106-46-7
Melting Point 52°C to 54°C (lit.)
Density 1.241 g/mL (at 25°C (literature))
Boiling Point 173°C (lit.)
Molecular Formula C6H4Cl2
Linear Formula C6H4Cl2
MDL Number MFCD00000604
Quantity 100 g
Molecular Weight (g/mol) 147
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.