Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Diethynylbenzene

Catalog No. 6320905G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-209-05G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-209-05G Supplier Sigma Aldrich Supplier No. 6320905G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triethyl orthopropionate is commonly used in the claisen rearrangement to construct new c-c bonds
  • It can react with pyrrole and chloroacetic acid to form 1,1,1- [tri-(pyrrol-2-yl)]propane

Specifications

CAS 935-14-8
Melting Point 94°C to 98°C (lit.)
Molecular Formula C10H6
Linear Formula HC///CC6H4C///CH
Quantity 100 mL
MDL Number MFCD00078375
Molecular Weight (g/mol) 126.15
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.