Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diethynylbenzene, 95%

Catalog No. p-3757669
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391550010 1 g Glass bottle
AC391550050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391550010 Supplier Thermo Scientific Chemicals Supplier No. 391550010
Only null left
Add to Cart

CAS: 935-14-8 | C10H6 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-14-8
Molecular Formula C10H6
Molecular Weight (g/mol) 126.16
MDL Number MFCD00078375
InChI Key MVLGANVFCMOJHR-UHFFFAOYSA-N
Synonym p-diethynylbenzene, benzene, 1,4-diethynyl, 1,4-diethynyl-benzene, benzene, diethynyl, benzene,1,4-diethynyl, benzene, p-diethynyl-7ci,8ci, 1,4-bis ethynyl benzene, acmc-209rm8, 4-05-00-01805 beilstein handbook reference
PubChem CID 120463
IUPAC Name 1,4-diethynylbenzene
SMILES C#CC1=CC=C(C=C1)C#C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 94°C to 98°C
Color Brown-Orange or White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(C∽CH)2
Quantity 1 g
Formula Weight 126.16
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4-Diethynylbenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.