Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diethynylbenzene, 97%, Thermo Scientific™

Catalog No. AAB2173103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2173103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2173103 Supplier Thermo Scientific Chemicals Supplier No. B2173103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-14-8
Molecular Formula C10H6
Molecular Weight (g/mol) 126.158
MDL Number MFCD00078375
InChI Key MVLGANVFCMOJHR-UHFFFAOYSA-N
Synonym p-diethynylbenzene, benzene, 1,4-diethynyl, 1,4-diethynyl-benzene, benzene, diethynyl, benzene,1,4-diethynyl, benzene, p-diethynyl-7ci,8ci, 1,4-bis ethynyl benzene, acmc-209rm8, 4-05-00-01805 beilstein handbook reference
PubChem CID 120463
IUPAC Name 1,4-diethynylbenzene
SMILES C#CC1=CC=C(C=C1)C#C

Specifications

Melting Point 94°C to 96°C
Quantity 1 g
UN Number UN1325
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material 1,4-Diethynylbenzene
Hazard Category Warning
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.