Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Difluoro-2-nitrobenzene, 98%, Thermo Scientific™

Catalog No. AAA1180722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1180722 100 g
AAA1180714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1180722 Supplier Thermo Scientific Chemicals Supplier No. A1180722

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-74-9
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00007054
InChI Key XNJAYQHWXYJBBD-UHFFFAOYSA-N
Synonym 2,5-difluoronitrobenzene, benzene, 1,4-difluoro-2-nitro, 1,4-difluoro-2-nitro-benzene, pubchem2289, 2,5 difluoronitrobenzene, 2.5-difluoronitrobenzene, acmc-1cjb9, 2,5-difluoro-nitrobenzene, 2,5-difluoronitro-benzene
PubChem CID 67767
IUPAC Name 1,4-difluoro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=CC=C1F

Specifications

Melting Point -12°C
Density 1.462
Boiling Point 206°C to 207°C
Flash Point 90°C (194°F)
Refractive Index 1.511
Quantity 100 g
UN Number UN2810
Beilstein 2210200
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 1,4-Difluoro-2-nitrobenzene
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-663-4
RTECSNumber CZ5715000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.