Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Difluorobenzene, 99+%

Catalog No. ac114610250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114610250 25 mL Glass bottle
AC114611000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC114610250 Supplier Thermo Scientific Chemicals Supplier No. 114610250
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 540-36-3 | C6H4F2 | 114.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-36-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000344
InChI Key QUGUFLJIAFISSW-UHFFFAOYSA-N
Synonym p-difluorobenzene, benzene, 1,4-difluoro, para-difluorobenzene, benzene, p-difluoro, 2,5-difluorobenzene, unii-y9fq2ef0z6, 1,4-difluorbenzol, y9fq2ef0z6, benzene,4-difluoro, pubchem2309
PubChem CID 10892
ChEBI CHEBI:38585
IUPAC Name 1,4-difluorobenzene
SMILES FC1=CC=C(F)C=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -13°C
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 88°C to 89°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4400 to 1.442
Linear Formula C6H4F2
Quantity 25 mL
Beilstein 05, 199
Specific Gravity 1.17
Merck Index 15, 3167
Formula Weight 114.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 4-Difluorobenzene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-742-9
RTECSNumber CZ5658000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.