Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylic acid, 3,5-diethyl ester | 632-93-9 | 99.5% | 267.32 g/mol | C14H21NO4 | 5 G
SDP

Catalog No. 5000226377
Encompass_Preferred

Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate is a research-grade small-molecule standard (CAS 632-93-9) used in biochemical and toxicology studies, including liver disease models. Supplied as a white to off-white powder, it is suitable for analytical assays and model development where high-purity pyridinedicarboxylate esters are required.

  • Chemical formula C14H21NO4; molecular weight 267.32 g/mol.
  • High purity (HPLC) 99.5%.
  • Low water content (KF) 0.02%.
  • Physical state: white to off-white powder.
  • Storage: powder stable -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Common use: research models of cholestatic liver injury and related studies.

Catalog No. 50-002-26377 Supplier Medchemexpress LLC Supplier No. HY1185945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate is a research-grade small-molecule standard (CAS 632-93-9) used in biochemical and toxicology studies, including liver disease models. Supplied as a white to off-white powder, it is suitable for analytical assays and model development where high-purity pyridinedicarboxylate esters are required.

  • Chemical formula C14H21NO4; molecular weight 267.32 g/mol.
  • High purity (HPLC) 99.5%.
  • Low water content (KF) 0.02%.
  • Physical state: white to off-white powder.
  • Storage: powder stable -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Common use: research models of cholestatic liver injury and related studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.