Learn More
Medchemexpress LLC 1,4-dihydroxy-2-naphthoic acid | 31519-22-9 | MFCD00010370 | 99.6% | 204.18 | C11H8O4 | 1 ML
SDP
Catalog No. 5000303789
Encompass_Preferred
1,4-Dihydroxy-2-naphthoic acid is a small-molecule aryl hydrocarbon receptor (AhR) agonist and a bacterially derived metabolite reported to exhibit anti-inflammatory activity. It is provided for research use as analytical standards and solution formulations for biological assays and analytical applications.
  • Acts as an aryl hydrocarbon receptor (AhR) agonist.
  • Reported to have anti-inflammatory activity in biological models.
  • Available as 10 mM solution in DMSO and as solid powder.
  • Supplied with high purity suitable for analytical and assay use.
  • Intended for use as an analytical standard and reagent in biochemical assays.
Catalog No. 50-003-03789 Supplier Medchemexpress LLC Supplier No. HYW01470110MM/1ML
1,4-Dihydroxy-2-naphthoic acid is a small-molecule aryl hydrocarbon receptor (AhR) agonist and a bacterially derived metabolite reported to exhibit anti-inflammatory activity. It is provided for research use as analytical standards and solution formulations for biological assays and analytical applications.
  • Acts as an aryl hydrocarbon receptor (AhR) agonist.
  • Reported to have anti-inflammatory activity in biological models.
  • Available as 10 mM solution in DMSO and as solid powder.
  • Supplied with high purity suitable for analytical and assay use.
  • Intended for use as an analytical standard and reagent in biochemical assays.