Learn More
Medchemexpress LLC 1,4-dihydroxy-2-naphthoic acid | 31519-22-9 | MFCD00010370 | 99.6% | 204.18 g/mol | C11H8O4 | 1 G
SDP
Catalog No. 5000302723
Encompass_Preferred
1,4-Dihydroxy-2-naphthoic acid is a small-molecule aryl hydrocarbon receptor (AhR) agonist and a bacterially derived metabolite reported to have anti-inflammatory activity. Supplied for research use, it is available in solid and ready-to-use solution formats, with supporting analytical and handling documents for laboratory studies.
  • Acts as an aryl hydrocarbon receptor (AhR) agonist
  • Is a bacterially derived metabolite with reported anti-inflammatory activity
  • High purity suitable for biochemical and pharmacological research
  • Available as a solid and as a 10 mM solution in DMSO
  • Includes analytical documentation such as COA, HNMR, and LCMS
Catalog No. 50-003-02723 Supplier Medchemexpress LLC Supplier No. HYW0147011G
1,4-Dihydroxy-2-naphthoic acid is a small-molecule aryl hydrocarbon receptor (AhR) agonist and a bacterially derived metabolite reported to have anti-inflammatory activity. Supplied for research use, it is available in solid and ready-to-use solution formats, with supporting analytical and handling documents for laboratory studies.
  • Acts as an aryl hydrocarbon receptor (AhR) agonist
  • Is a bacterially derived metabolite with reported anti-inflammatory activity
  • High purity suitable for biochemical and pharmacological research
  • Available as a solid and as a 10 mM solution in DMSO
  • Includes analytical documentation such as COA, HNMR, and LCMS