Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dihydroxy-2-naphthoic Acid 98.0+%, TCI America™
SDP

Catalog No. D229625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D229625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D229625G Supplier TCI America Supplier No. D229625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31519-22-9
Molecular Formula C11H8O4
Molecular Weight (g/mol) 204.18
MDL Number MFCD00010370
InChI Key VOJUXHHACRXLTD-UHFFFAOYSA-N
Synonym 1,4-dihydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalenecarboxylic acid, 1,4-dihydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 1,4-dihydroxy, unii-1u77mr5q7o, 1,4-dihydroxy-2-naphtoic acid, 1,4-dohydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalic acid, cbmicro_022187, acmc-209hn7
PubChem CID 671
ChEBI CHEBI:18094
IUPAC Name 1,4-dihydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=CC(O)=C2C=CC=CC2=C1O

Specifications

Melting Point 191°C
Quantity 25 g
Formula Weight 204.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dihydroxy-2-naphthoic Acid
EINECSNumber (4)-1468
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.