Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dimethoxy-2-nitrobenzene 99.0+%, TCI America™
SDP

Catalog No. D063825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D063825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D063825G Supplier TCI America Supplier No. D063825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-39-4
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.163
MDL Number MFCD00024211
InChI Key UPTOWXNJLZJTGD-UHFFFAOYSA-N
Synonym Nitrodimethylhydroquinone, 2-Nitrohydroquinone Dimethyl Ether
PubChem CID 66639
IUPAC Name 1,4-dimethoxy-2-nitrobenzene
SMILES COC1=CC(=C(C=C1)OC)[N+](=O)[O-]

Specifications

Melting Point 72°C
Boiling Point 169°C
Quantity 25 g
Formula Weight 183.16
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dimethoxy-2-nitrobenzene
EINECSNumber (3)-0801
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.