Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Dimethoxybenzene

Catalog No. D1313501KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1313501KG 1 kg
D131350500G 500 g
D131350100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1313501KG Supplier Sigma Aldrich Supplier No. D1313501KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Dimethoxybenzene

Specifications

CAS 150-78-7
Melting Point 54°C to 56°C (lit.)
Density 1.053 g/mL (at 25°C (literature))
Boiling Point 213°C (lit.)
Molecular Formula C8H10O2
Linear Formula C6H4(OCH3)2
MDL Number MFCD00008401
Quantity 1 kg
Synonym p-Methoxyanisole; Dimethylhydroquinone; Hydroquinone dimethyl ether; NSC 7483
Molecular Weight (g/mol) 138.16
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.