Learn More
Medchemexpress LLC 1,4-dimethoxybenzene | 150-78-7 | MFCD00008401 | 100.0% | 138.16 g/mol | C8H10O2 | 1 KG
SDP
Catalog No. 5000300519
Encompass_Preferred
1,4-Dimethoxybenzene (C8H10O2) is an aromatic ether used as an organic reagent and reference standard in chemical research. It is supplied at very high purity for synthesis, analytical calibration, and method development.
  • High purity 99.99% suitable for analytical and synthetic applications.
  • Characterized by H NMR and GC, with COA and SDS available.
  • Available in multiple sizes, including 1 kg, for bulk use.
  • Also available as 10 mM solutions in DMSO for ready-to-use applications.
  • Useful as a reagent, reference standard, and research chemical.
  • Molecular weight 138.16 g/mol; molecular formula C8H10O2.
Catalog No. 50-003-00519 Supplier Medchemexpress LLC Supplier No. HYW0157801KG
1,4-Dimethoxybenzene (C8H10O2) is an aromatic ether used as an organic reagent and reference standard in chemical research. It is supplied at very high purity for synthesis, analytical calibration, and method development.
  • High purity 99.99% suitable for analytical and synthetic applications.
  • Characterized by H NMR and GC, with COA and SDS available.
  • Available in multiple sizes, including 1 kg, for bulk use.
  • Also available as 10 mM solutions in DMSO for ready-to-use applications.
  • Useful as a reagent, reference standard, and research chemical.
  • Molecular weight 138.16 g/mol; molecular formula C8H10O2.