Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-dimethoxybenzene | 150-78-7 | MFCD00008401 | 100.0% | 138.16 g/mol | C8H10O2 | 100 G
SDP

Catalog No. 5000300751
Encompass_Preferred

1,4-Dimethoxybenzene is an analytical reference standard (CAS 150-78-7) intended for research and analytical applications. It is a white to off-white crystalline solid (C8H10O2, 138.16 g/mol) commonly used as a reference in qualitative and quantitative HPLC, GC, and MS methods. Purity is typically 99.99%; storage and handling should follow the certificate of analysis to preserve sample integrity.

  • High purity (99.99%) for reliable analytical results.
  • Suitable as a reference standard for HPLC, GC, and MS methods.
  • White to off-white solid form for easy handling and weighing.
  • Chemical formula C8H10O2 and molecular weight 138.16 g/mol for unambiguous identification.
  • Storage and shipping guidance provided in the certificate of analysis to maintain integrity.

Catalog No. 50-003-00751 Supplier Medchemexpress LLC Supplier No. HYW015780100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-Dimethoxybenzene is an analytical reference standard (CAS 150-78-7) intended for research and analytical applications. It is a white to off-white crystalline solid (C8H10O2, 138.16 g/mol) commonly used as a reference in qualitative and quantitative HPLC, GC, and MS methods. Purity is typically 99.99%; storage and handling should follow the certificate of analysis to preserve sample integrity.

  • High purity (99.99%) for reliable analytical results.
  • Suitable as a reference standard for HPLC, GC, and MS methods.
  • White to off-white solid form for easy handling and weighing.
  • Chemical formula C8H10O2 and molecular weight 138.16 g/mol for unambiguous identification.
  • Storage and shipping guidance provided in the certificate of analysis to maintain integrity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.