Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-dimethoxybenzene | 150-78-7 | MFCD00008401 | 100.0% | 138.16 g/mol | C8H10O2 | 25 MG
SDP

Catalog No. 5000303421
Encompass_Preferred

1,4-Dimethoxybenzene (CAS 150-78-7) analytical standard supplied as a high-purity reference material for research and analytical applications. Offered in small-scale quantities suitable for method development, calibration, and qualitative or quantitative analysis.

  • High purity 99.99% for accurate analytical results.
  • Available in small quantities, including 25 mg, for lab-scale use.
  • Supplied as a solid reference standard for chromatography and spectroscopy.
  • Molecular weight 138.16 g/mol and chemical formula C8H10O2 for documentation.
  • SMILES provided to support informatics and structure confirmation.

Catalog No. 50-003-03421 Supplier Medchemexpress LLC Supplier No. 25MGHYW015780R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-Dimethoxybenzene (CAS 150-78-7) analytical standard supplied as a high-purity reference material for research and analytical applications. Offered in small-scale quantities suitable for method development, calibration, and qualitative or quantitative analysis.

  • High purity 99.99% for accurate analytical results.
  • Available in small quantities, including 25 mg, for lab-scale use.
  • Supplied as a solid reference standard for chromatography and spectroscopy.
  • Molecular weight 138.16 g/mol and chemical formula C8H10O2 for documentation.
  • SMILES provided to support informatics and structure confirmation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.