Learn More
Medchemexpress LLC 1,4-dimethoxybenzene | 150-78-7 | MFCD00008401 | 100.0% | 138.16 g/mol | C8H10O2 | 50 MG
SDP
Catalog No. 5000300060
Encompass_Preferred
Analytical reference standard of 1,4-dimethoxybenzene (CAS 150-78-7) supplied as a high-purity crystalline solid for research and analytical applications. It is intended for instrument calibration, method development, and quality control where a well-characterized reference compound is required.
  • High purity (99.99%) for analytical reliability.
  • Molecular formula C8H10O2, molecular weight 138.16 g/mol.
  • White to colorless crystalline solid with melting point ~55-60 °C.
  • Suitable for calibration, method development, and quality control.
  • Provided in small, measured quantities for laboratory use.
Catalog No. 50-003-00060 Supplier Medchemexpress LLC Supplier No. HYW015780R50MG
Analytical reference standard of 1,4-dimethoxybenzene (CAS 150-78-7) supplied as a high-purity crystalline solid for research and analytical applications. It is intended for instrument calibration, method development, and quality control where a well-characterized reference compound is required.
  • High purity (99.99%) for analytical reliability.
  • Molecular formula C8H10O2, molecular weight 138.16 g/mol.
  • White to colorless crystalline solid with melting point ~55-60 °C.
  • Suitable for calibration, method development, and quality control.
  • Provided in small, measured quantities for laboratory use.