Learn More
Medchemexpress LLC 1,4-dimethoxybenzene | 150-78-7 | MFCD00008401 | 100.0% | 138.17 g/mol | C8H10O2 | 1 ML
SDP
Catalog No. 5000300518
Encompass_Preferred
1,4-dimethoxybenzene (CAS 150-78-7) is an endogenous metabolite provided for research and analytical use. The product is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid sizes, with reported purity of 99.99%. Preclinical notes include anxiolytic-like effects in animal studies and a negative in vivo micronucleus test for clastogenicity.
  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • High reported purity: 99.99%.
  • Suitable for research and analytical applications.
  • Also available in multiple solid sizes for bulk use.
  • Documented preclinical biological data and non-clastogenic in vivo micronucleus result.
Catalog No. 50-003-00518 Supplier Medchemexpress LLC Supplier No. HYW01578010MM/1ML
1,4-dimethoxybenzene (CAS 150-78-7) is an endogenous metabolite provided for research and analytical use. The product is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and in multiple solid sizes, with reported purity of 99.99%. Preclinical notes include anxiolytic-like effects in animal studies and a negative in vivo micronucleus test for clastogenicity.
  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • High reported purity: 99.99%.
  • Suitable for research and analytical applications.
  • Also available in multiple solid sizes for bulk use.
  • Documented preclinical biological data and non-clastogenic in vivo micronucleus result.