Learn More
Medchemexpress LLC 1,4-dimethoxybenzene-d10 | 74079-00-8 | 98.0% | 148.23 g/mol | C8D10O2 | 25 MG
SDP
Catalog No. 5000303388
Encompass_Preferred
1,4-Dimethoxybenzene-d10 is the deuterium-labeled form of 1,4-dimethoxybenzene used as an analytical standard and tracer in research. It is employed as an internal standard for NMR, GC-MS, and LC-MS applications, and in metabolic and aromatic profiling studies. The compound has formula C8D10O2 and high isotopic enrichment (about 98 atom % D).
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS
  • High isotopic enrichment for reliable quantitation
  • Suitable tracer for metabolic and aromatic profiling studies
  • Stable solid form for convenient storage and handling
  • Compatible with common analytical solvents and methods
Catalog No. 50-003-03388 Supplier Medchemexpress LLC Supplier No. 25MGHYW015780S
1,4-Dimethoxybenzene-d10 is the deuterium-labeled form of 1,4-dimethoxybenzene used as an analytical standard and tracer in research. It is employed as an internal standard for NMR, GC-MS, and LC-MS applications, and in metabolic and aromatic profiling studies. The compound has formula C8D10O2 and high isotopic enrichment (about 98 atom % D).
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS
  • High isotopic enrichment for reliable quantitation
  • Suitable tracer for metabolic and aromatic profiling studies
  • Stable solid form for convenient storage and handling
  • Compatible with common analytical solvents and methods