Learn More
Medchemexpress LLC 1,4-dimethoxybenzene-d10 | 74079-00-8 | 99.6% | 148.23 | C8D10O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015780S100MG
Encompass_Preferred
1,4-Dimethoxybenzene-d10 is the deuterium-labeled analogue of 1,4-dimethoxybenzene, supplied as a crystalline reference standard for use as an internal standard and tracer in analytical chemistry and metabolomics. It is characterized by high isotopic enrichment and is provided in small lab-scale pack sizes for GC-MS and LC-MS workflows.
  • Deuterium-labeled reference standard for analytical and metabolomics studies.
  • High isotopic purity (reported purity 99.6%).
  • Molecular formula C8D10O2 and molecular weight 148.23 Da.
  • White to off-white crystalline appearance for storage and handling.
  • Available in small pack sizes suitable for analytical use.
Catalog No. 50-003-00691