Learn More
Medchemexpress LLC 1,4-dimethoxybenzene-d10 | 74079-00-8 | 99.6% | 148.23 | C8D10O2 | 50 MG

Supplier: Medchemexpress LLC HYW015780S50MG
1,4-Dimethoxybenzene-d10 is the deuterium-labeled analogue of 1,4-dimethoxybenzene used as a stable isotope tracer and reference standard in laboratory research. It is supplied as a crystalline, high-purity material intended for analytical applications and metabolic studies.
- Deuterium-labeled tracer and reference standard for analytical studies.
- High isotopic enrichment and 99.6% purity.
- Crystalline, white to off-white solid form.
- Molecular formula C8D10O2 and molecular weight 148.23.
- Suitable for GC-MS and metabolic tracer experiments.
- Recommended storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
- Supplied in a 50 mg pack size for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.