Learn More
Medchemexpress LLC 1,4-dimethoxybenzene (standard) | 150-78-7 | MFCD00008401 | 100.0% | 138.16 g/mol | C8H10O2 | 250 MG
SDP
Catalog No. 5000302361
Encompass_Preferred
1,4-dimethoxybenzene (standard) is an analytical reference material of 1,4-dimethoxybenzene (CAS 150-78-7) intended for research and analytical applications. It is supplied at very high purity for use in method development, calibration, and quantitative analysis in laboratory settings.
  • Provides high-purity reference material for analytical assays.
  • Suitable for chromatography and mass spectrometry calibration.
  • Available in multiple pack sizes to suit laboratory needs.
  • Supplied with safety data documentation for safe handling and use.
  • Intended for research, method development, and quality control applications.
Catalog No. 50-003-02361 Supplier Medchemexpress LLC Supplier No. 250MGHYW015780R
1,4-dimethoxybenzene (standard) is an analytical reference material of 1,4-dimethoxybenzene (CAS 150-78-7) intended for research and analytical applications. It is supplied at very high purity for use in method development, calibration, and quantitative analysis in laboratory settings.
  • Provides high-purity reference material for analytical assays.
  • Suitable for chromatography and mass spectrometry calibration.
  • Available in multiple pack sizes to suit laboratory needs.
  • Supplied with safety data documentation for safe handling and use.
  • Intended for research, method development, and quality control applications.