Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dimethylcyclohexane (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. D0700500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0700500ML 500 mL
D070025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0700500ML Supplier TCI America Supplier No. D0700500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 589-90-2
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00001508,MFCD00064174,MFCD00064955
InChI Key QRMPKOFEUHIBNM-UHFFFAOYSA-N
Synonym Hexahydro-p-xylene
PubChem CID 11523
IUPAC Name 1,4-dimethylcyclohexane
SMILES CC1CCC(C)CC1

Specifications

Color Colorless
Quantity 500 mL
UN Number 2263
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dimethylcyclohexane (cis- and trans- mixture)
EINECSNumber (3)-2227
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.