Learn More
ATHENA CONTROLS INC 1/4 DIN TEMPERATURE CONTROLLER
Catalog No. NC3591050
Encompass
  • 1/4 DIN panel mounted form factor.
  • Universal temperature and process controller.
  • Auto-tuning PID and on/off control modes.
  • Single loop control.
  • Supports input calibration for thermocouples, RTD, current linear, and voltage.
  • Output 1 is a normally open relay.
  • Output 2 is not applicable.
  • Alarm 1 is a normally open relay.
  • Alarm 2 is a normally open relay.
  • No communication capabilities.
  • Dual digital LED display type.
  • Power supply operates at 115/230 VAC, 50/60 Hz.
  • Compliance with CE standards.
Catalog No. NC3591050 Supplier ATHENA CONTROLS INC Supplier No. 25CAB0BB0000CE
  • 1/4 DIN panel mounted form factor.
  • Universal temperature and process controller.
  • Auto-tuning PID and on/off control modes.
  • Single loop control.
  • Supports input calibration for thermocouples, RTD, current linear, and voltage.
  • Output 1 is a normally open relay.
  • Output 2 is not applicable.
  • Alarm 1 is a normally open relay.
  • Alarm 2 is a normally open relay.
  • No communication capabilities.
  • Dual digital LED display type.
  • Power supply operates at 115/230 VAC, 50/60 Hz.
  • Compliance with CE standards.