Learn More
AA BLOCKS LLC 1,4-DIOXA-SPIRO(4,5)DEC-7-25G
Supplier:  AA BLOCKS LLC AA0038F025G
Encompass_Preferred
1,4-DIOXA-SPIRO[4,5]DEC-7-EN-8-BORONIC ACID, PINACOL ESTER | Purity: 98% | Mol Wt: 266.1410 | 680596-79-6 | MFCD04115656 | 25 g
Catalog No. NC1935350