Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187636 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9915 5 mL
11-101-9077 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9915 Supplier MilliporeSigma Supelco Supplier No. 768875MLF
Only null left
Add to Cart
Add to Cart

1,4-Dioxane is an organic compound and an environmental contaminant, generally found in industrial waste streams and by-products. It is commonly used as an industrial solvent, in cosmetic products and household products such as lotions, soap, shampoos, bath foams.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 356°F
CAS 123-91-1
Vapor Pressure 27 mmHg (20°C)
Vapor Density 3 (vs. air)
Density 1.034 g/mL (at 25°C)
Flash Point 12°C
Physical Form Neat
Linear Formula C4H8O2
MDL Number MFCD00006571
Melting Point 10°C to 12°C
Boiling Point 100°C to 102°C
Synonym Diethylene oxide; Dioxane
Type Analytical Standard
Quantity 5 mL
Formula Weight 88.11
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.