Learn More
Thermo Scientific™ 1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles
Catalog No. AA47305M1
Change view
Click to view available options
Quantity:
100 mL
1 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47305M1 4 x 1 L
AA47305AE 100 mL
AA47305K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA47305M1 Supplier Thermo Scientific™ Supplier No. 47305M1