Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane (p-Dioxane; Diethylene Ether), Reagents

Catalog No. 01337277
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-277 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-277 Supplier Reagents Holdings Llc Supplier No. C2197000500C
 Restricted Item
Add to Cart
Add to Cart

1,4-Dioxane (p-Dioxane; Diethylene Ether), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 123-91-1
Boiling Range 101.2°C
Melting Point 12°C
Color Colorless
Boiling Point 101.2°C
Melting Range 12°C
Packaging Glass Bottle
Molecular Formula C4H8O2
Quantity 500 mL
Specific Gravity 1.034
Synonym p-Dioxane, 1,4-Diethylene dioxide
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES C1COCCO1
IUPAC Name 1,4-dioxane
Molecular Weight (g/mol) 88.11
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,4-Dioxane (p-Dioxane, Diethylene Ether)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.