Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,4-Dioxane solution NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm x 8 in. | 123-91-1 | MFCD00006571 | 1EA
Catalog No. 501882563
Encompass
1,4-Dioxane solution NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm x 8 in. | Mol Wt: 88.11 | 123-91-1 | MFCD00006571 | 1EA
Catalog No. 50-188-2563 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6842281EA
1,4-Dioxane solution NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm x 8 in. | Mol Wt: 88.11 | 123-91-1 | MFCD00006571 | 1EA