Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,4-Dioxane solution NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm x 8 in. | 123-91-1 | MFCD00006571 | 1EA
Catalog No. 501882555
Encompass
1,4-Dioxane solution NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm x 8 in. | Mol Wt: 88.11 | 123-91-1 | MFCD00006571 | 1EA
Catalog No. 50-188-2555 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6119051EA
Add to Cart
Add to Cart
1,4-Dioxane solution NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm x 8 in. | Mol Wt: 88.11 | 123-91-1 | MFCD00006571 | 1EA