Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diphenyl-1,3-butadiene, Scintillationgrade 99+%, Thermo Scientific™

Catalog No. AC409350250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC409350250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC409350250 Supplier Thermo Scientific Chemicals Supplier No. 409350250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for fluorescence/luminescence spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-65-7
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00004791
InChI Key JFLKFZNIIQFQBS-FNCQTZNRSA-N
Synonym 1e,3z-1,4-diphenyl-1,3-butadiene, cis,trans-1,4-diphenylbutadiene, e,z-1,4-diphenyl-1,3-butadiene, cis,trans-1,4-diphenyl-1,3-butadiene, 1,3-butadiene, 1,4-diphenyl-, e,z, 1,4-diphenyl-1,3-butadiene, 1e,3z, benzene,1'-1,3-butadiene-1,4-diyl bis, 1z,3e-4-phenylbuta-1,3-dienyl benzene, benzene,1,1'-1,3-butadiene-1,4-diyl bis, 1,4-diphenyl-1,3-butadiene e,z-form mi
PubChem CID 1551381
IUPAC Name [(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
SMILES C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula C6H5CH=CHCH=CHC6H5
Quantity 25 g
Beilstein 05, 676
Fieser 01,337
Merck Index 15, 3350
Formula Weight 206.29
Percent Purity ≥99.0%
Physical Form Crystals or Needles
Chemical Name or Material 1, 4-Diphenyl-1, 3-butadiene, 99+%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.