Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diphenyl-1-butanone, 98%

Catalog No. AAL1559703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1559703 1 g
AAL1559706 5 g
AAL1559714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1559703 Supplier Thermo Scientific Chemicals Supplier No. L1559703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5407-91-0 | C16H16O | 224.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5407-91-0
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.303
MDL Number MFCD00022005
InChI Key GBUMEGLMTNAXOM-UHFFFAOYSA-N
PubChem CID 79413
IUPAC Name 1,4-diphenylbutan-1-one
SMILES C1=CC=C(C=C1)CCCC(=O)C2=CC=CC=C2

Specifications

Melting Point 54°C to 56°C
Boiling Point 180°C (8 mmHg)
Quantity 1 g
Synonym 4-phenylbutyrophenone, 1,4-diphenyl-1-butanone, 1-butanone, 1,4-diphenyl, 4-phenylbutyrophenon, pubchem8921, 1-butanone,4-diphenyl, 1,4-diphenyl-butan-1-one, 1-butanone,1,4-diphenyl, 3-phenylpropyl phenyl ketone, acmc-1am65
Formula Weight 224.3
Percent Purity 98%
Chemical Name or Material 1,4-Diphenyl-1-butanone
EINECSNumber 226-471-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.