Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diphenyl-3-thiosemicarbazide 99.0+%, TCI America™
SDP

Catalog No. D102625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D102625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D102625G Supplier TCI America Supplier No. D102625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1768-59-8
Molecular Formula C13H13N3S
Molecular Weight (g/mol) 243.328
MDL Number MFCD00022107
InChI Key VGIVYSWGHVFQRP-UHFFFAOYSA-N
PubChem CID 775364
IUPAC Name 1-anilino-3-phenylthiourea
SMILES C1=CC=C(C=C1)NC(=S)NNC2=CC=CC=C2

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 25 g
Formula Weight 243.33
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diphenyl-3-thiosemicarbazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.