Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diphenylsemicarbazide 98.0+%, TCI America™
SDP

Catalog No. D091325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D091325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D091325G Supplier TCI America Supplier No. D091325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 621-12-5
Molecular Formula C13H13N3O
Molecular Weight (g/mol) 227.267
MDL Number MFCD00020635
InChI Key NGZZNUMYERKSQA-UHFFFAOYSA-N
PubChem CID 69299
IUPAC Name 1-anilino-3-phenylurea
SMILES C1=CC=C(C=C1)NC(=O)NNC2=CC=CC=C2

Specifications

Melting Point 181°C
Color White
Quantity 25 g
Formula Weight 227.27
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diphenylsemicarbazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.