Learn More
Cayman Chemical 1,4-DPCA ethyl ester, Purity: ≥98%, CAS: 86443-19-8, 50 mg
SDP
Catalog No. 502916310
Encompass_Preferred
A synthetic precursor of 1,4-DPCA; active against M. tuberculosis (MIC90 = 23.99 µM)
Catalog No. 50-291-6310 Supplier Cayman Chemical Supplier No. 3833450 MG
A synthetic precursor of 1,4-DPCA; active against M. tuberculosis (MIC90 = 23.99 µM)